Learn More
Corning™ BTD Volume Rocker Switch
Corning™ BTD volume rocker switch
165.20 SEK - 192.89 SEK
Specifications
Description | BTD volume rocker switch |
---|---|
For Use With (Application) | Dispensing Organic or Inorganic Liquids |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
16882100
|
Corning™
6859 |
2.5 to 50 mL |
165.20 SEK
Each |
Estimated Shipment: 03-12-2024
Log in to see stock. |
|||||
16892100
|
Corning™
6860 |
100 mL |
192.89 SEK
Each |
Estimated Shipment: 03-12-2024
Log in to see stock. |
|||||
Specifications
BTD volume rocker switch | |
Dispensing Organic or Inorganic Liquids |