Learn More
Hettich Lab Technology™ Bucket
Bucket comes in a variety of designs to accommodate customized centrifuge loads.
Brand: Hettich Lab Technology™ 1745
This item is not returnable.
View return policy
Description
- Available in a variety of configurations
- Configurations capable of accommodating a host of customized centrifuge workloads
Specifications
2 x 25 mL | |
Bucket | |
25 mL |
26.0 OD x 78 mm L | |
2 |