missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution in Dosing Bottle, ph 7.00, Testo™
Click to view available options
Quantity:
250 mL
Description
- pH buffer solution 7.00 in dosing bottle
- 250mL
Specifications
Specifications
pH | 7 |
Quantity | 250 mL |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'