missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution, pH 12.000, NIST, Chem-Lab
Click to view available options
Quantity:
10 L
Packaging:
PE
Description
pH = 12.58 at 0°
pH = 12.26 at 10°
pH = 12.00 at 20°
pH = 11.88 at 25°
pH = 11.75 at 30°
pH = 11.53 at 40°
pH = 11.31 at 50°
pH = 11.09 at 60°
pH = 10.88 at 70°
pH = 10.68 at 80°
pH = 10.48 at 90°C
pH = 12.26 at 10°
pH = 12.00 at 20°
pH = 11.88 at 25°
pH = 11.75 at 30°
pH = 11.53 at 40°
pH = 11.31 at 50°
pH = 11.09 at 60°
pH = 10.88 at 70°
pH = 10.68 at 80°
pH = 10.48 at 90°C
Specifications
Specifications
Density | 1.01g/mL |
pH | 12.00 |
Quantity | 10 L |
Formulation | Phosphate, NaOH |
Packaging | PE |
Traceability to NIST | NIST traceable |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Buffer Solution, pH 12.000, NIST, Chem-Lab >
Spot an opportunity for improvement?Share a Content Correction