Learn More
Brady™ Bulk Polyethylene Tags for M710, M610, M611
Bulk Polyethylene Tags for use with Brady™ Printers M611, M610 and M710.
Brand: Brady™ 312126
Additional Details : Weight : 0.65500kg
Description
- Multipurpose tag that can be used for a variety of applications including identification of multi-conductor cables, inventory, equipment, lockout, safety warning repair and work-in-progress
- Extremely tear resistant and ideal cold-weather tag
- Cross-laminated polyethylene provides high tear resistance and cold-weather performance
Specifications
100 Labels | |
ASTM D 1000, ASTM D 1938, ASTM G 115, ASTM G 155, RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC | |
Polyethylene | |
Data and Telecommunications Labeling, Wire and Cable Labeling;Inventory and inspection labelling;Lockout tagout;Safety identification;Wire and Cable Labeling | |
1.97 in. | |
0.216 mm | |
-40°F to +120°F | |
12 mm | |
5.59 x 1.58 mm |
Tag | |
White | |
Matte | |
50 mm | |
Rectangle | |
-40°C to +49°C | |
0.472 in. | |
0.22 x 0.062 in. | |
M610, M710, M611, BMP61, BMP71 |