Learn More
C. botulinum BoNT-A Light Chain Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Brand: Invitrogen PA548053
3104.50 SEK valid until 2025-03-16
Use promo code "24070" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant C. botulinum BoNTA Heavy Chain is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
C. botulinum BoNT-A Light Chain Polyclonal antibody specifically detects C. botulinum BoNT-A Light Chain in Bacteria samples. It is validated for Immunoprecipitation, Western Blot.
Specifications
C. botulinum BoNT-A Light Chain | |
Polyclonal | |
Unconjugated | |
BONTC1 | |
E. coli-derived recombinant Clostridium BoNT-A Light Chain Met1-Phe425 | |
100 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Sheep | |
Antigen affinity chromatography | |
RUO | |
Bacteria | |
Antibody | |