missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C19orf70 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | C19orf70 |
---|---|
Concentration | 0.25 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16824085
|
Proteintech
25514-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16814085
|
Proteintech
25514-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody is specific to C19orf70.
Component of the MICOS complex, a large protein complex of the mitochondrial inner membrane that plays crucial roles in the maintece of crista junctions, inner membrane architecture, and formation of contact sites to the outer membrane.Specifications
C19orf70 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
Q5XKP0 | |
125988, 224904 | |
C19orf70 Fusion Protein Ag22003 | |
Primary | |
-20°C | |
MICOS13 |
0.25 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C19orf70, MIC13, P117, Protein P117, Protein QIL1, QIL1 | |
Micos13 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |