Learn More
Uvex™ C500 Dry Cut Protection Gloves
Click to view available options
Size:
7
8
9
10
Description
- Bamboo TwinFlex™ Technology ensures a very high level of cut protection
- HPV dots on the palm for enhanced grip capabilities
- Highly flexible with good tactile sensation
- Uvex Climazone effectively removes excess warmth and moisture
- Free of harmful substances in accordance with OEKO-TEX™ Standard 100
- Silicone-free according to the imprint test
- 3D Ergo Technology ensures a secure fit
Specifications
Coating Material | HPV Dots |
Color | Green, Black |
Material | Bamboo Viscose, HPPE, Polyamide |
Size | 8 |
EN388 Standard | XX4XC |
Glove Cuff Style | Knit |
Series | C500 Dry |