missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Cabinet Colorstack Slide
Brand: Epredia™ E43/12C/LILAC
Additional Details : Weight : 12.00000kg
Description
14 drawer holds 76mm x 26mm microscope slides vertically 6000 slide capacity lilac Epredia™ Raymond LambSpecifications
Lilac | |
6000 Slides | |
Slide Storage | |
48 cm |
Cabinets with Drawers | |
48 x 48 x 14 cm | |
14 cm | |
48 cm |