missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Calcitonin (8-32), Salmon
For cellular and molecular biology applications
821.26 SEK - 1160.00 SEK
Specifications
Species | Salmon |
---|---|
Purification Method | HPLC |
Content And Storage | -30°C to -10°C |
Format | Solid |
Molecular Weight (g/mol) | 2725.06 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15476369
|
Thermo Scientific™
J64415.|D |
0.5 mg |
821.26 SEK
0.50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15486369
|
Thermo Scientific™
J64415.M¬ |
1 mg | |||||||
Description
Specifications
Salmon | |
-30°C to -10°C | |
2725.06 | |
RUO - research use only |
HPLC | |
Solid | |
Calcitonin (8-32), salmon | |
Val-Leu-Gly-Lys-Leu-Ser-Gln-Glu-Leu-His-Lys-Leu-Gln-Thr-Tyr-Pro-Arg-Thr-Asn-Thr-Gly-Ser-Gly-Thr-Pro-NH2 |
RUO – Research Use Only