Learn More
Calcium dihydrogen phosphate hydrate, 97%, Thermo Scientific Chemicals
593.00 SEK - 6165.00 SEK
Chemical Identifiers
CAS | 301524-28-7 |
---|---|
Molecular Formula | CaH4O8P2 |
Molecular Weight (g/mol) | 234.05 |
MDL Number | MFCD00010898 |
InChI Key | YYRMJZQKEFZXMX-UHFFFAOYSA-L |
Synonym | Calcium phosphate, monobasic; Calcium tetrahydrogen diphosphate |
IUPAC Name | calcium didihydrogen phosphate |
SMILES | [Ca++].OP(O)([O-])=O.OP(O)([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11480063
|
Thermo Scientific Alfa Aesar
A12123.36 |
500 g |
593.00 SEK
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11490063
|
Thermo Scientific Alfa Aesar
A12123.0E |
2500 g |
2065.00 SEK
2500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11400073
|
Thermo Scientific Alfa Aesar
A12123.0C |
10,000 g |
6165.00 SEK
10000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Calcium dihydrogen phosphate hydrate is a pH control reagent, stabilizer for plastics, manufacture of glass.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCalcium dihydrogen phosphate hydrate is a pH control reagent, stabilizer for plastics, manufacture of glass.
Solubility
Soluble in water 18 g/L @ 30°C.
Notes
Loses H{2}O at 100°, dec. at 200°
Chemical Identifiers
301524-28-7 | |
234.05 | |
YYRMJZQKEFZXMX-UHFFFAOYSA-L | |
calcium didihydrogen phosphate |
CaH4O8P2 | |
MFCD00010898 | |
Calcium phosphate, monobasic; Calcium tetrahydrogen diphosphate | |
[Ca++].OP(O)([O-])=O.OP(O)([O-])=O |
Specifications
∼200°C (decomposition) | |
CaH4O8P2 | |
None | |
Calcium phosphate, monobasic; Calcium tetrahydrogen diphosphate | |
YYRMJZQKEFZXMX-UHFFFAOYSA-L | |
calcium didihydrogen phosphate | |
234.06 (Anhydrous) | |
Calcium dihydrogen phosphate hydrate |
301524-28-7 | |
MFCD00010898 | |
14,1693 | |
Soluble in water 18g/L @ 30°C. | |
[Ca++].OP(O)([O-])=O.OP(O)([O-])=O | |
234.05 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 231-837-1
missing translation for 'rtecsNumber' : TB8527000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Calcium dihydrogen phosphate hydrate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Phosphoric acid, calcium salt
RUO – Research Use Only