missing translation for 'onlineSavingsMsg'
Learn More

Calcium L-threonate, 98%, Thermo Scientific Chemicals

Product Code. 15457579
Click to view available options
Quantity:
100 g
250 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Calcium L-threonate is reactant involved in synthesis of cyclic hydroxamates and substituted oxazinanones and oxidative degradation to yield oxalate complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Calcium L-threonate is reactant involved in synthesis of cyclic hydroxamates and substituted oxazinanones and oxidative degradation to yield oxalate complexes.

Solubility
Soluble in water. Sparingly soluble in methanol.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70753-61-6
Molecular Formula C8H14CaO10
Molecular Weight (g/mol) 310.27
MDL Number MFCD00077882,MFCD00150824
InChI Key ZJXGOFZGZFVRHK-UHFFFAOYNA-L
Synonym calcium l-threonate, l-threonic acid calcium salt, calcium 2r,3s-2,3,4-trihydroxybutanoate, calcium threonate, unii-hbb4ypo93u, hbb4ypo93u, calcium bis l-threonate, threonine, calcium salt, bis l-threonic acid calcium salt, butanoic acid, 2,3,4-trihydroxy-, calcium salt 2:1 , 2r,3s
PubChem CID 13388558
IUPAC Name calcium;(2R,3S)-2,3,4-trihydroxybutanoate
SMILES [Ca++].OCC(O)C(O)C([O-])=O.OCC(O)C(O)C([O-])=O

Specifications

Melting Point >300°C
Color Brown
Quantity 250 g
Solubility Information Soluble in water. Sparingly soluble in methanol.
Formula Weight 310.27
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Calcium L-threonate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.