Learn More
Hanna Instruments™ Calcium Reagents
Reagents for the colorimetric determination of calcium using a compatible benchtop or portable photometer.
Brand:  Hanna Instruments™ HI937521-01
Product Code. 12209329
  • 943.00 SEK / Pack of 100
Estimated Shipment: 19-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Specifications
100
0 to 400mg/L
Calcium Reagent
100/Pk.
Oxalate
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hanna Instruments™ Calcium Reagents > Method: Oxalate; Number of Tests: 100; Range: 0 to 400mg/L