missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Sulfate Dihydrate, 98.0-102.0%, (Crystalline/ACS Reagent Grade), MP Biomedicals
253.00 SEK - 2015.00 SEK
Chemical Identifiers
CAS | 10101-41-4 |
---|---|
Molecular Formula | CaH4O6S |
Molecular Weight (g/mol) | 172.16 |
MDL Number | MFCD00149625 |
InChI Key | PASHVRUKOFIRIK-UHFFFAOYSA-L |
Synonym | calcium sulfate dihydrate, phosphogypsum, landplaster, annaline, compactrol, gips, gypsite, primoplast, satinite, gypsum stone |
PubChem CID | 24928 |
ChEBI | CHEBI:32583 |
IUPAC Name | calcium dihydrate sulfate |
SMILES | O.O.[Ca++].[O-]S([O-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11440512
|
MP Biomedicals
0219141480 |
100 g |
253.00 SEK
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11450512
|
MP Biomedicals
0219141490 |
500 g |
1160.00 SEK
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11460512
|
MP Biomedicals
0219141491 |
1 kg |
2015.00 SEK
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
10101-41-4 | |
172.16 | |
PASHVRUKOFIRIK-UHFFFAOYSA-L | |
24928 | |
calcium dihydrate sulfate |
CaH4O6S | |
MFCD00149625 | |
calcium sulfate dihydrate, phosphogypsum, landplaster, annaline, compactrol, gips, gypsite, primoplast, satinite, gypsum stone | |
CHEBI:32583 | |
O.O.[Ca++].[O-]S([O-])(=O)=O |
Specifications
10101-41-4 | |
98.0-102.0% | |
MFCD00149625 | |
PASHVRUKOFIRIK-UHFFFAOYSA-L | |
calcium dihydrate sulfate | |
24928 | |
172.2 | |
Calcium Sulfate Dihydrate |
Crystalline | |
CaH4O6S | |
calcium sulfate dihydrate, phosphogypsum, landplaster, annaline, compactrol, gips, gypsite, primoplast, satinite, gypsum stone | |
O.O.[Ca++].[O-]S([O-])(=O)=O | |
172.16 | |
CHEBI:32583 | |
ACS Reagent |
Safety and Handling
Recommended Storage : Room Temperature