missing translation for 'onlineSavingsMsg'
Learn More

Calretinin Mouse anti-Human, Mouse, Pig, Rat, CoraLite 555, Clone: 2D7A9, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech CL555-66496-20UL

 View more versions of this product

Product Code. 17781359

  • 1645.54 SEK / 20µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Calretinin Monoclonal antibody apecifically detetects Calretinin antigen in Human, Mouse, Pig, Rat Samples. It is validated for Immunofluorescence.
Specifications

Specifications

Calretinin
Monoclonal
1000 ug/mL
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3.
CALB2
Calretinin fusion protein Ag2924
20 μL
794
Store at -20°C. Avoid exposure to light.
IgG1
Immunofluorescence
2D7A9
CoraLite 555
BC015484
Mouse
Protein G purification
Primary
Human, Mouse, Pig, Rat
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers