missing translation for 'onlineSavingsMsg'
Learn More
Camptothecin, 97.48%, MP Biomedicals™ Product Code.: 11455811

Camptothecin, 97.48%, MP Biomedicals™

Product Code. 11455811 Shop All MP Biomedical Products
100 mg
Click to view available options
Quantity:
100 mg
This item is not returnable. View return policy

Product Code. 11455811

Brand: MP Biomedicals 0215973280

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 7691-02-3
Color Yellow
Assay Percent Range 0.9748
Molecular Formula C8H19NSi2
Quantity 100 mg
Synonym 1,1,3,3-tetramethyl-1,3-divinyldisilazane, 1,3-divinyltetramethyldisilazane, 1,3-divinyl-1,1,3,3-tetramethyldisilazane, silanamine, 1-ethenyl-n-ethenyldimethylsilyl-1,1-dimethyl, bis dimethyl vinyl silyl amine, ccris 2450, bis ethenyldimethylsilyl amine, ethenyl-dimethylsilyl amino-dimethylsilyl ethene, n-dimethylvinylsilyl-1,1-dimethyl-1-vinylsilylamine, dvtmds
InChI Key WYUIWUCVZCRTRH-UHFFFAOYSA-N
SMILES C[Si](C)(C=C)N[Si](C)(C)C=C
IUPAC Name [[[ethenyl(dimethyl)silyl]amino]-dimethylsilyl]ethene
Molecular Weight (g/mol) 185.42
PubChem CID 82126
Formula Weight 348.34
Physical Form Solid
Chemical Name or Material Camptothecin
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Camptothecin, 97.48%, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.