missing translation for 'onlineSavingsMsg'
Learn More
Camptothecin 97% MP Biomedicals Product Code.: 11435811

Camptothecin 97% MP Biomedicals

Product Code. 11435811 Shop All MP Biomedical Products
25 mg
Click to view available options
Quantity:
25 mg
50 mg
250 mg
This item is not returnable. View return policy

Product Code. 11435811

Brand: MP Biomedicals 0215973225

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 7689-03-4
Molecular Formula C20H16N2O4
Molecular Weight (g/mol) 348.36
MDL Number MFCD00081076
InChI Key VSJKWCGYPAHWDS-FQEVSTJZSA-N
Synonym camptothecin, camptothecine, s-+-camptothecin, campathecin, +-camptothecine, d-camptothecin, 20 s-camptothecine, +-camptothecin, 21,22-secocamptothecin-21-oic acid lactone, s-camptothecinShow MoreShow Less
PubChem CID 24360
ChEBI CHEBI:27656
SMILES CCC1(C2=C(COC1=O)C(=O)N3CC4=CC5=CC=CC=C5N=C4C3=C2)O

Specifications

Color Yellow
Density 1.51g/cm3
Assay Percent Range >97%
Quantity 25 mg
Formula Weight 348.3
Physical Form Solid
Chemical Name or Material Camptothecin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Camptothecin 97% MP Biomedicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.