missing translation for 'onlineSavingsMsg'
Learn More

Candida albicans Monoclonal Antibody (A97H), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen MA518176

Product Code. 15302657

  • 4545.00 SEK / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This antibody is reactive with all C. albicans strains tested (9), as well as C. glabrata and C. tropicales. It does not react with C. parapsilosis or C. krusei. Purity is >90%.

Candida albicans Monoclonal antibody specifically detects Candida albicans in Yeast samples. It is validated for ELISA, Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Candida albicans
Monoclonal
100 μg/mL
PBS with 0.1% sodium azide; pH 7.2
Mouse
Protein A
RUO
1 Each
Yeast
Liquid
ELISA, Immunocytochemistry
A97H
Unconjugated
C. albicans
Candida albicans native protein
100 μg
Primary
0.1% Sodium azide
4° C
IgM
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.