Learn More
Thermo Scientific™ Capitol Vial Collection Cup Holder
Thermo Scientific™ Capitol Vial Collection Cup and Cup Holder are used for for traditional and on-site testing.
Brand:  Thermo Scientific™ 98740
Additional Details : Weight : 0.95000kg
Product Code. 15628578
  • 217.00 SEK / Pack of 40
Estimated Shipment: 11-12-2024
to see stock.
Add to basket
Description
Description
Thermo Scientific™ Capitol Vial Collection Cup and Cup Holder are used for for traditional and on-site testing. The Capitol Vial Collection Cup Holder holds the 8 ounce collection cups, Cat #10126 and Cat #10122TS.
  • Material: Polypropylene
Specifications
Specifications
Collection Cup Holder
Polypropylene
Holder for 8 ounce Beaker
40/Cs.