Learn More
Thermo Scientific™ Capitol Vial Collection Cup Holder
Thermo Scientific™ Capitol Vial Collection Cup and Cup Holder are used for for traditional and on-site testing.
Brand: Thermo Scientific™ 98740
Additional Details : Weight : 0.95000kg
Description
Thermo Scientific™ Capitol Vial Collection Cup and Cup Holder are used for for traditional and on-site testing. The Capitol Vial Collection Cup Holder holds the 8 ounce collection cups, Cat #10126 and Cat #10122TS.
- Material: Polypropylene
Specifications
Collection Cup Holder | |
Polypropylene | |
Holder for 8 ounce Beaker | |
40/Cs. |