Learn More
Thermo Scientific™ Capitol Vial Dual Pocket Specimen Transport Bags
GREENER_CHOICE
Capitol Vial Dual Pocket Biohazard Bags for Specimen Transport
5585.00 SEK
Specifications
No. per Case 1000/Cs.
View More Specs
Products 1
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
17390240
GREENER_CHOICE
View Documents
Thermo Scientific™
20315
Dual Pocket Specimen Transport Bag with Absorbent Sheet
5585.00 SEK
Pack of 1000
Description
Description
Capitol Vial Specimen Transport Bags are ideal for the collection, storage and transport of biological specimens. The dual pocket design allows for specimens to be safely secured in the front pocket with an absorbent pad and chain of custody paperwork can be stowed in the rear pocket without risk of sample exposure.
  • Dual pocket bag for sample and documentation security
  • Features a sealing tamper evident sealing flap
  • Bags are printed Biohazard symbol and instructions for use
Ordering Information:
Custom printed biohazard bags available, minimum quantity required.
Includes:
Transport bag includes absorbent pad
Specifications
Specifications
1000/Cs.