missing translation for 'onlineSavingsMsg'
Learn More

(Carbethoxymethyl)triphenylphosphonium bromide, 98%, Thermo Scientific Chemicals

Product Code. 10666482
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Specifications

CAS 1530-45-6
CAS Min % 97.5
CAS Max % 100.0
Melting Point 158.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C22H22BrO2P
Linear Formula C2H5O2CCH2P(C6H5)3Br
MDL Number MFCD00011835
Quantity 25 g
Synonym carbethoxymethyl triphenylphosphonium bromide, 2-ethoxy-2-oxoethyl triphenylphosphonium bromide, ethoxycarbonylmethyl triphenylphosphonium bromide, carbethoxymethylenetriphenylphosphonium bromide, ethoxycarbonylmethyl triphenyl phosphonium bromide, phosphonium, 2-ethoxy-2-oxoethyl triphenyl-, bromide, carbethoxymethyl-triphenyl-phosphonium bromide, 2-ethoxy-2-oxoethyl triphenylphosphanium bromide, carboethoxymethyl triphenylphosphonium bromide, phosphonium, ethoxycarbonylmethyl triphenyl-, bromide
InChI Key VJVZPTPOYCJFNI-UHFFFAOYSA-M
SMILES [Br-].CCOC(=O)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name (2-ethoxy-2-oxoethyl)-triphenylphosphanium;bromide
Molecular Weight (g/mol) 429.29
PubChem CID 73731
Formula Weight 429.28
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material (Carbethoxymethyl)triphenylphosphonium bromide, 98%
Show More Show Less
compliance-icons
Product Identifier
  • (Carbethoxymethyl)triphenylphosphonium bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'