Learn More
Carboxypeptidase A4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand:  Invitrogen PA5122164
 View more versions of this product
Product Code. 17723564
  • 4180.00 SEK / 100µL
Explore more special offers
This item is not returnable. View return policy
Description
Description
Carboxypeptidase A4 is a member of the carboxypeptidase A/B subfamily. Carboxypeptidases are zinc-containing exopeptidases that catalyze the release of carboxy-terminal amino acids, and are synthesized as zymogens that are activated by proteolytic cleavage. This protein could be involved in the histone hyperacetylation pathway. It is imprinted and may be a strong candidate protein for prostate cancer aggressiveness.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Carboxypeptidase A4
Polyclonal
Unconjugated
CPA4
1110019K20Rik; AV009555; Carboxypeptidase A3; carboxypeptidase A4; CPA3; Cpa4; RGD1563619; UNQ694/PRO1339
Rabbit
Antigen affinity chromatography
RUO
51200
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300
Q9UI42
CPA4
Lys55-Tyr421 (Accession Q9UI42), with N-terminal His Tag
100 μL