Learn More
Carboxypeptidase A4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5122164
This item is not returnable.
View return policy
Description
Carboxypeptidase A4 is a member of the carboxypeptidase A/B subfamily. Carboxypeptidases are zinc-containing exopeptidases that catalyze the release of carboxy-terminal amino acids, and are synthesized as zymogens that are activated by proteolytic cleavage. This protein could be involved in the histone hyperacetylation pathway. It is imprinted and may be a strong candidate protein for prostate cancer aggressiveness.![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Carboxypeptidase A4 | |
Polyclonal | |
Unconjugated | |
CPA4 | |
1110019K20Rik; AV009555; Carboxypeptidase A3; carboxypeptidase A4; CPA3; Cpa4; RGD1563619; UNQ694/PRO1339 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51200 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q9UI42 | |
CPA4 | |
Lys55-Tyr421 (Accession Q9UI42), with N-terminal His Tag | |
100 μL | |
Primary | |
Human | |
Antibody | |