Learn More
STEMCELL Technologies CAY10512
NF-κB pathway inhibitor; Inhibits NF-κB
Brand:  STEMCELL Technologies 73022
Additional Details : Weight : 0.05000kg
Product Code. 17140162
  • POA / 100mg
Estimated Shipment: 20-09-2024
to see stock.
This item requires a price update prior to being ordered.
Request a Price for this item here.
Description
Description
  • CAY10512 is an analog of Resveratrol that is 100-fold more potent (IC50 = 150 nM) at inhibiting NF-κB activation by TNF-α (Heynekamp et al.) Unlike Resveratrol and some other trans-stilbene analogs, CAY10512 does not exhibit antioxidant activity (up to 15 μM) in either the ferric reducing/antioxidant power (FRAP) or total radical antioxidant parameter (TRAP) assays (Heynekamp et al.) Like Resveratrol, CAY10512 also inhibits lipopolysaccharide-induced expression of COX-2 (Heynekamp et al.)
  • Protects human islets from instant blood-mediated inflammatory response leading to improved survival of transplanted islets in in vitro tube model (Kanak et al.)
  • Inhibits induction of NF-κB and subsequent upregulation of inflammatory microRNAs in Alzheimer disease model of human neuronal-glial cells in vitro (Lukiw)
Specifications
Specifications
139141-12-1
100 mg
C15H13FO
≥97%