missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR5 Rabbit anti-Human, Clone: RC-10, Novus Biologicals™
Rabbit Monoclonal Antibody
2455.00 SEK - 5410.00 SEK
Specifications
Antigen | CCR5 |
---|---|
Clone | RC-10 |
Dilution | Western Blot, ELISA, Immunocytochemistry/Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18361362
|
Bio-Techne
NBP3-12000-0.025MG |
0.025 mg |
2455.00 SEK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18337544
|
Bio-Techne
NBP3-12000-0.2MG |
0.2 mg |
5410.00 SEK
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCR5 Monoclonal antibody specifically detects CCR5 in Human samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
CCR5 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
C-C CKR-5, C-C motif chemokine receptor 5 A159A, CCCKR5, CC-CKR-5FLJ78003, CCR-5, CD195, CD195 antigen, chemokine (C-C motif) receptor 5, chemokine receptor CCR5, chemr13, CKR5, CKR-5, HIV-1 fusion coreceptor, IDDM22CMKBR5C-C chemokine receptor type 5 | |
This antibody was raised by immunizing mice with CCR5. | |
Primary |
RC-10 | |
Monoclonal | |
Rabbit | |
Alzheimers Research, Cancer, Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Neurodegeneration, Neuroscience, Phospho Specific, Signal Transduction | |
PBS | |
1234 | |
IgG κ | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title