missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD235a Monoclonal Antibody (JC159), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA512484
This item is not returnable.
View return policy
Description
CD235a (Glycophorins A, GPA) is a single pass membrane sialoglycoprotein expressed in mature erythrocytes and erythroid precursor cells. CD235a is the carrier of blood group M and N specificities. CD235a provides cells with a large mucin-like that may serve as a barrier to cell fusion, minimizing aggregation between red blood cells in the circulation. CD235a has been shown to act as a receptor for Sandei virus, parvovirus, and Hsa, an Streptococcus adhesin.Specifications
CD235a | |
Monoclonal | |
0.2 mg/mL | |
10mM PBS with 0.05% BSA and 0.05% sodium azide, pH 7.4 | |
P02724 | |
GYPA | |
Immunogen Membrane preparation from splenic hairy cell leukemia | |
500 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG1 κ |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
JC159 | |
Unconjugated | |
GYPA | |
AI853584; CD235; CD235a; erythroid-lineage-specific membrane sialoglycoprotein; glycophorin A; glycophorin A (MN blood group); glycophorin A (MNS blood group); glycophorin A, GPA; glycophorin Erik; glycophorin MiI; glycophorin MiV; glycophorin SAT; glycophorin Sta type C; glycophorin-A; GPA; GPErik; GPSAT; Gypa; HGNC:4702; HGpMiV; HGpMiX; HGpMiXI; HGpSta(C); HGpStaC; Mi.V glycoprotein; Mi.V glycoprotein (24 AA); MN; MN sialoglycoprotein; MNS; PAS-2; recombinant glycophorin A-B Miltenberger-DR; sialoglycoprotein alpha | |
Mouse | |
Protein A/G | |
RUO | |
2993, 688972 | |
4° C | |
Liquid |