Learn More
CD24 Mouse anti-Human, Unconjugated, Clone: 1H5C4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67627-1-IG-150UL
This item is not returnable.
View return policy
Description
The CD24 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD24. This antibody recognizes human antigen. The CD24 antibody has been validated for the following applications: FC, WB, ELISA analysis.
Specifications
CD24 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Flow Cytometry, ELISA | |
1H5C4 | |
Unconjugated | |
150 μL | |
100133941 | |
IgG1 |