missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD24 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA549555
This item is not returnable.
View return policy
Description
CD24 is a two-chain glycosylphosphatidylinositol (GPI)-anchored glycoprotein expressed at multiple stages of B-cell development, beginning with the bone marrow pro-B-cell compartment and continuing through mature, surface Ig positive B-cells. Plasma cell expression is very low or negative. CD24 is also expressed on the majority of B-lineage acute lymphoblastic leukemias, B-cell CCLs and B-cell non-Hodgkin′s lymphomas. CD24 may play a role in regulation of B-cell proliferation and maturation, and control of autoimmunity.Specifications
Cd24 | |
Polyclonal | |
Unconjugated | |
CD24 | |
Signal transducer CD24, Heat-stable antigen, HSA, Nectadrin, CD24, Cd24, Cd24a | |
Rabbit | |
Protein A | |
RUO | |
100133941, 12484, 25145 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
5 mg/mL | |
TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
P25063, P24807, Q07490 | |
CD24, Cd24a | |
synthetic peptide corresponding to 27-53aa of rat CD24 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |