missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD38, Rabbit anti-Human, Clone: AT13/5, Novus Biologicals™
Rabbit Monoclonal Antibody
2535.00 SEK - 5610.00 SEK
Specifications
Antigen | CD38 |
---|---|
Clone | AT13/5 |
Concentration | 1 mg/mL |
Dilution | Flow Cytometry |
Applications | Flow Cytometry |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18272349
|
Bio-Techne
NBP2-81055-0.025MG |
0.025 mg |
2535.00 SEK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18331970
|
Bio-Techne
NBP2-81055-0.2MG |
0.2 mg |
5610.00 SEK
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD38 Monoclonal specifically detects CD38 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
CD38 | |
1 mg/mL | |
Flow Cytometry | |
Unconjugated | |
Rabbit | |
Apoptosis, B Cell Development and Differentiation Markers, Cell Biology, Hematopoietic Stem Cell Markers, Immunology, Stem Cell Markers | |
PBS | |
952 | |
Namalwa cells emulsified in CFA. | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
AT13/5 | |
Flow Cytometry | |
Monoclonal | |
Purified | |
RUO | |
Human | |
ADP-ribosyl cyclase 1, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase, cADPr hydrolase 1, CD38 antigen, CD38 antigen (p45), CD38 molecule, Cyclic ADP-ribose hydrolase 1, EC 3.2.2.5, NAD(+) nucleosidase, T10 | |
CD38 | |
IgG κ | |
Protein A purified | |
Binds specifically to human CD38, an approximately 45 kDa type II transmembrane protein, expressed on essentially all pre-B lymphocytes, plasma cells, and thymocytes. Also present on activated T lymphocytes, natural killer (NK) lymphocytes, myeloblasts, and erythroblasts. Bimodally expressed during B cell development, modulating from high in immature cells to low in intermediate ones and back to high on mature B cells. This antibody competes with clone HB7 (Ab00128) (Ellis 1995) which has been shon to bind to an epitope between amino acids 273-285). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title