missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD79a Monoclonal Antibody (ZL7-4), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA181093
This item is not returnable.
View return policy
Description
MA1-81093 reacts with human CD79a-positive cells by flow cytometry and ELISA specific for a fusion protein of CD79a-Fc. This clone has been reported to be useful in distinguishing B-CLL from mantle cell lymphoma in flow cytometric assays. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human CD79a antibody, clone ZL7-4 recognizes the human B-cell antigen receptor complex-associated protein alpha chain, also known as MB-1 membrane glycoprotein or CD79a.
CD79a is a disulphide-linked heterodimer that includes B29 (CD79b) polypeptide. CD79a is a B lymphocyte antigen receptor with an antigen-specific surface component Ig (immunoglobulin) that associates with Ig-alpha and Ig-beta, necessary elements for the expression and function of the B-cell antigen receptor. CD79a first appears at pre-B cell stage and persists until the plasma cell stage where it is found as an intracellular component. CD79a is found in the majority of acute leukemias of precursor B cell type, in B cell lines, B cell lymphomas, and in some myelomas. It is not present in myeloid or T cell lines. Diseases associated with CD79a include Agammaglobulinemia 3 and Agammaglobulinemia, Non-Bruton type.Specifications
CD79a | |
Monoclonal | |
1 mg/mL | |
TBS with 0.09% sodium azide, pH 7.4 | |
P11912 | |
CD79A | |
IgM complex isolated from Daudi cells. | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation | |
ZL7-4 | |
Unconjugated | |
CD79A | |
B-cell antigen receptor; B-cell antigen receptor complex-associated protein alpha chain; B-cell antigen receptor complex-associated protein alpha chain-like; CD79; CD79a; CD79A antigen (immunoglobulin-associated alpha); CD79a molecule; CD79a molecule, immunoglobulin-associated alpha; CD79A protein; CD79-alpha protein; EGK_10665; Ig alpha; IGA; Igalpha; ig-alpha; immunoglobulin-associated alpha; LOC100913063; Ly54; Ly-54; MB1; MB-1; MB-1 membrane glycoprotein; Membrane-bound immunoglobulin-associated protein; surface IgM-associated protein | |
Mouse | |
Protein A | |
RUO | |
973 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |