missing translation for 'onlineSavingsMsg'
Learn More

CD95 Hamster anti-Mouse, PE, Clone: JO2, BD

Hamster Monoclonal Antibody

Brand:  BD Biosciences 554258

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15877628

  • 5165.00 SEK / Each
Estimated Shipment: 29-08-2024
to see stock.

Description

Description

Fas antigen, CD95, is a 45kDa cell-surface protein which can mediate apoptosis. It belongs to the TNF (tumor necrosis factor)/NGF receptor family. Expression of Fas has been described in the thymus, liver, heart, lung and ovary. Fas plays an important role in the apoptotic process that takes place during development. Monoclonal antibodies recognizing Fas such as Jo2 have cytolytic activity on cells expressing Fas. The cell death stimulated by Fas antibodies is characteristic of apoptosis and suggests that the lethal effects are a result of interaction of antibody with a functional Fas antigen as opposed to complement-mediated lysis.
The Jo2 antibody recognizes mouse Fas. The Jo2 antibody shows cytolytic activity against cell lines expressing mouse Fas by inducing apoptosis. Intraperitoneal injections of Jo2 mAb have been shown to kill mice and induce apoptotic hepatocyte death. Jo2 mAb immunoprecipitates mouse Fas as a 45kDa band from W4 cells. W4 cells are WR19L mouse lymphoma cells transformed with mouse Fas. The difference between the observed MW of Fas and that deduced from its amino acid sequence (Mr 34,971) may be due to glycosylation.

Host Species: Hamster
Clone: Jo2
Isotype: IgG2 λ2
Species Reactivity: Mouse
Immunogen: WR19L mouse lymphoma cells transformed with recombinant mouse Fas

Flow Cytometry

Specifications

Specifications

CD95
Monoclonal
0.2mg/mL
Apo-1; Apt1; Fas; FASLG receptor; lpr; TNFR6; Tnfrsf6; TNR6
Hamster
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
JO2
PE
Aqueous buffered solution containing ≤0.09% sodium azide.
WR19L mouse lymphoma cells transformed with recombinant mouse Fas
0.2 mg
Immunology
Mouse
IgG2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.