missing translation for 'onlineSavingsMsg'
Learn More

Cefadroxil, MP Biomedicals™

Product Code. 11427881
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 50370-12-2
Molecular Formula C16H17N3O5S
Molecular Weight (g/mol) 363.388
MDL Number MFCD00865091
InChI Key BOEGTKLJZSQCCD-UEKVPHQBSA-N
Synonym p-hydroxycephalexine, Cefamox
PubChem CID 47965
ChEBI CHEBI:3479
IUPAC Name (6R,7R)-7-[[(2R)-2-amino-2-(4-hydroxyphenyl)acetyl]amino]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
SMILES CC1=C(N2C(C(C2=O)NC(=O)C(C3=CC=C(C=C3)O)N)SC1)C(=O)O

Specifications

Type Cefadroxil
Physical Form Crystalline Powder
Formula Weight 363.4
Color White
Quantity 1 g
Chemical Name or Material Cefadroxil
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'