Learn More
Ceftazidime, MedChemExpress
MedChemExpress Ceftazidime (GR20263), an antibiotic, has a broad spectrum activity against Gram-positive and Gram-negative aerobic bacteria. Ceftazidime is also active against Enterobacteriaceae (including β-lactamase-positive strains) and is resistant to hydrolysis by most β-lactamases.
879.15 SEK - 1326.17 SEK
Specifications
Chemical Name or Material | Ceftazidime |
---|---|
CAS | 72558-82-8 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C22H22N6O7S2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16402249
|
Medchem Express
HY-B0593 |
1 g |
879.15 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16461018
|
Medchem Express
HY-B0593 |
5 g |
1326.17 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ceftazidime (GR20263), an antibiotic, has a broad spectrum activity against Gram-positive and Gram-negative aerobic bacteria. Ceftazidime is also active against Enterobacteriaceae (including β-lactamase-positive strains) and is resistant to hydrolysis by most β-lactamases.Specifications
Ceftazidime | |
White | |
C22H22N6O7S2 | |
H2O : 100 mg/mL (182.96 mM; Need ultrasonic) ∣DMSO : 25 mg/mL (45.74 mM; ultrasonic and warming and heat to 80°C) | |
546.58 | |
98.0% | |
Powder |
72558-82-8 | |
Research | |
GR20263 | |
O=C1[C@@H](NC(/C(C2=CSC(N)=N2)=N\OC(C)(C(O)=O)C)=O)[C@@]3([H])SCC(C[N+]4=CC=CC=C4)=C(C([O-])=O)N13 | |
546.58 | |
Research | |
COVID-19-immunoregulation |
Safety and Handling
H317∣H319∣H334
missing translation for 'shelfLife' : 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Recommended Storage : 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.