missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Celite™ 512 Medium, Calcined Filter Aid, Honeywell Fluka™
Click to view available options
Quantity:
1 kg
Packaging:
Poly Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Color | Brown to White |
Quantity | 1 kg |
CAS | 91053-39-3 |
Packaging | Poly Bottle |
Molecular Formula | O2Si |
MDL Number | MFCD00132803 |
UN Number | NONH for all modes of transport |
InChI Key | VYPSYNLAJGMNEJ-UHFFFAOYSA-N |
SMILES | O=[Si]=O |
IUPAC Name | dioxosilane |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'