missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ Cell Line Nucleofector™ Kit V
Use for high transfection efficiency with siRNA duplexes even in suspension cells. Cell Line Nucleofector™ Kit V enables expression within hours - from transfection to analysis in a day.
Brand: Lonza™ VCA-1003
This item is not returnable.
View return policy
Description
- Includes:
- Nucleofector SolutionV
- Supplement
- pmax GFP Vector
- Certified cuvettes
- Plastic cuvettes
- For Use With Nucleofector Device

Specifications
4°C | |
Achieve transfection efficiencies of up to 90% with high cell viability | |
Get up to 99% transfection efficiency with siRNA duplexes even in suspension cells, Expression within hours - from transfection to analysis in a day | |
Cell Line Nucleofector Kit V |
pmaxGFP™ | |
Nucleofector™ Device | |
25 |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lonza™ Cell Line Nucleofector™ Kit V > Number of Reactions: 25
Spot an opportunity for improvement?Share a Content Correction