Learn More
STEMCELL Technologies CellAdhere™ Type I Collagen, Human, Lyophilized
Type I collagen, human, for tissue engineering research, cell culture, and biochemistry
Brand:  STEMCELL Technologies 07005
Additional Details : Weight : 0.01500kg
Product Code. 15202755
  • 9529.65 SEK / 15mg
Estimated Shipment: 04-09-2024
to see stock.
Description
Description
  • Type I Collagen is the most abundant type of collagen in the human body and is a major structural protein prevalent in skin, bone, tendon, and other fibrous connective tissues
  • All collagens are characterized by a secondary protein structure consisting of three polypeptide chains wrapped around one another in a triple helical arrangement
  • The different types of collagen vary in the primary amino acid sequence of their polypeptide chains, with type I collagen comprised of two identical α1(I) chains and one α2(I) chain that spontaneously form the triple helix in solution
  • This product is ideal for coating surfaces with a thin layer of protein to support cell attachment, but is not suitable for 3D cell culture applications
  • The optimal final concentration for coating may vary depending on the cell type, and therefore must be titrated for best results
Specifications
Specifications
15 mg