Learn More
Thermo Scientific™ CellSensor™ HRE-bla ME-180 Cell Line
The CellSensor™ HRE-bla ME-180 cell line contains a beta-lactamase reporter gene under control of the hypoxia response element (HRE) stably integrated into ME-180 cells.
Brand:  Thermo Scientific™ K1644
Product Code. 10266643
  • 143800.00 SEK / Each
Estimated Shipment: 16-12-2024
to see stock.
Explore more special offers
Add to basket
For Research Use Only. All usage must comply with product instructions.
This item is not returnable. View return policy
Description
Description
The CellSensor™ HRE-bla ME-180 cell line contains a beta-lactamase reporter gene under control of the hypoxia response element (HRE) stably integrated into ME-180 cells. To construct this cell line, the HRE-bla construct was transduced into ME-180 cells by lentivirus. Flow cytometry was used to isolate cells responsive to deferoxamine. This cell line is validated for DMSO tolerance, cell number, stimulation time, substrate loading time, as well as Z' and EC50 concentrations for cobalt chloride and deferoxamine. The CellSensor™ HRE-bla ME-180 cell line is responsive to deferoxamine and cobalt chloride and can be used to probe the hypoxia signaling pathway. This cell line can be adapted for high-throughput screening of agonist or antagonist compound libraries. Candidate drugs can also be tested for dose response against this cell line. Academic and non-profit customers, please inquire for special pricing.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cell-based beta lactamase reporter gene
CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.
CellSensor™
HIF1 alpha, Hypoxia pathway
CellSensor™
Dividing Cells
Signaling Pathway
1 Vial
Dividing Cells
Signaling Pathway
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Research Use Only. Not for use in diagnostic procedures.