Learn More
Thermo Scientific™ CellSensor™ NFAT-bla Jurkat Cell Line
The CellSensor™ NFAT-bla Jurkat cell line contains a beta-lactamase reporter gene under control of a Nuclear Factor of Activated T Cells (NFAT) response element stably integrated into Jurkat cells.
Brand: Thermo Scientific™ K1671B
Description
The CellSensor™ NFAT-bla Jurkat cell line contains a beta-lactamase reporter gene under control of a Nuclear Factor of Activated T Cells (NFAT) response element stably integrated into Jurkat cells. This cell line can be used as a parental cell line to build specific G-protein coupled receptor (GPCR) assays after transfection of additional genes of interest. NFAT-bla Jurkat cells have been shown to be responsive to phorbol 12-myristate 13-acetate (PMA) and Ionomycin stimulation. Additional testing data using alternate stimuli is available upon request. Academic and non-profit customers, please inquire for special pricing.
Specifications
Dividing Cells | |
CellSensor™ NFAT-bla Jurkat Cell Line, 1Vial, ∽ 20,000,000 cells/vial at 20,000,000 cells/mL Quantity of Cells, NFAT-bla Jurkat Cell Line, Fluorescent Detection Method, CellSensor™ Druggable Target, Suspension Growth Properties, Liquid Nitrogen Storage | |
Pharma and biopharma, drug discovery and development, target and lead identification and validation, pathway biology, cellular pathway analysis assays, cellsensor™ cellular pathway assays | |
1 vial | |
Cell Line | |
CellSensor™ |
CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. | |
Fluorescent | |
beta-lactamase reporter gene under control of a nuclear factor of activated T cells (NFAT) response element stably integrated into Jurkat cells | |
NFAT-bla Jurkat | |
Jurkat | |
Signaling Pathway |
Product Suggestions
Customers who viewed this item also viewed.