Learn More
Greiner Bio-One CELLSTAR CELLreactor™ Conical Bottom Polypropylene Filter Top Tube
For shaking suspension cell, aerobic bacteria and microorganisms.
Brand:  Greiner Bio-One 227245
Additional Details : Weight : 0.50000kg
Product Code. 12656147
  • 11228.89 SEK / Pack of 500
Estimated Shipment: 02-08-2024
to see stock.
Description
Description
  • Can be used as a small bioreactor for suspension and spheroid cell culture
  • Facilitates high number of parallel experiments
  • Flexible working volume from 1 to 35mL
  • Sterile
  • Allows gas exchange
Specifications
Specifications
Culture Tube
Free of detectable DNase/RNase, human DNA and pyrogens; non-cytotoxic
Polypropylene
Shaking suspension cell, aerobic bacteria and microorganisms
50 mL
Yes
Blue
Filter Screw Cap
Yes
Sterile
Conical Bottom