Learn More
Tocris Bioscience™ Cetrorelix Acetate
Potent GnRH receptor antagonist
Brand: Tocris Bioscience™ 3536/1
1885.76 SEK valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
Cetrorelix Acetate is a potent gonadotropin-releasing hormone (GnRH) receptor antagonist (KD = 0.202 nM, IC50 = 1.21 nM). Suppresses production of luteinizing hormone (LH) and follicle-stimulating hormone (FSH) from the pituitary gland, which inhibits ovulation. Exhibits antiproliferative effects and displays efficacy against hormone-sensitive cancers in vivo. Also exhibits anxiolytic and antidepressant activity in vivo.Specifications
Cetrorelix Acetate | |
Cetrorelix Acetate | |
Ac-D-2-Nal-D-(4-Cl-Phe)-D-(3-Pyridyl-Ala)-Ser-Tyr-D-Cit-Leu-Arg-Pro-D-Ala-NH2.CH3CO2H |
1491.11 | |
1 mg |
For Research Use Only