missing translation for 'onlineSavingsMsg'
Learn More

CGP 37849, Tocris Bioscience™

Product Code. 18095552
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective NMDA antagonist

Potent, selective and competitive NMDA receptor antagonist (Ki = 35 nM). Anticonvulsive following oral administration in vivo.

Chemical Identifiers

CAS 127910-31-0
Molecular Formula C6H12NO5P
Molecular Weight (g/mol) 209.138
InChI Key BDYHNCZIGYIOGJ-DUXPYHPUSA-N
Synonym 4-methyl-appa, e-2-amino-4-methyl-5-phosphonopent-3-enoic acid, 2-amino-4-methyl-5-phosphono-3-pentenoic acid, 3-pentenoic acid, 2-amino-4-methyl-5-phosphono, 3e-2-amino-4-methyl-5-phosphonopent-3-enoic acid, chembl29811, cgp hplc, e-2-amino-4-methyl-5-phosphono-3-pentenoic acid, e-+/--2-amino-4-methyl-5-phosphono-3-pentenoic acid, 3-pentenoic acid, 2-amino-4-methyl-5-phosphono-, e-+-
PubChem CID 5950212
IUPAC Name (E)-2-amino-4-methyl-5-phosphonopent-3-enoic acid
SMILES CC(=CC(C(=O)O)N)CP(=O)(O)O

Specifications

Quantity 50 mg
Formula Weight 209.14
Percent Purity >99%
Chemical Name or Material CGP 37849
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'