missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Chemical Test Kit

Includes all of the reagents and equipment necessary to perform manual water testing. Hanna Instruments™ Chemical Test Kit features pre-made, pre-measured reagents for fast, accurate determinations with high-resolution readings based on accepted titrations methods.

Brand:  Hanna Instruments™ HI38074

 View more versions of this product

Product Code. 11642340

  • 3830.00 SEK / Each
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

These chemical test kits use high quality reagents that are manufactured in Hanna's state-of-the-art facility and are clearly marked with the lot number and expiration date on each packet/bottle for traceability.

  • Complete setup: All required materials are included, such as beakers, syringes, and premeasured reagent bottles and packets
  • High resolution readings for fast, accurate determinations
  • Replacement reagents available: No need to buy a new kit when reagents are exhausted
Specifications

Specifications

Boric acid
Water samples
100
0.0 to 5.0mg/L
Titration
Boron determination
Reagents for 100 tests, HI98103 Checker pH meter, pH 4.01 sachet (1), pH 7.01 sachet (1), screwdriver, 120mL bottle with cap, 50mL calibrated vessel, and 1mL plastic pipettes (2)
1 Kit
Irrigation water testing
Chemical Test Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.