Learn More
Hanna Instruments™ Chemical Test Kit
Includes all of the reagents and equipment necessary to perform manual water testing. Hanna Instruments™ Chemical Test Kit features pre-made, pre-measured reagents for fast, accurate determinations with high-resolution readings based on accepted titrations methods.
Brand:  Hanna Instruments™ HI38001
 View more versions of this product
Product Code. 11622300
  • 3070.00 SEK / Each
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
These chemical test kits use high quality reagents that are manufactured in Hanna's state-of-the-art facility and are clearly marked with the lot number and expiration date on each packet/bottle for traceability.
  • Complete setup: All required materials are included, such as beakers, syringes, and premeasured reagent bottles and packets
  • High resolution readings for fast, accurate determinations
  • Replacement reagents available: No need to buy a new kit when reagents are exhausted
Specifications
Specifications
Barium chloride
Water samples
100 ea. (200 total)
100 to 1000mg/L and 1,000 to 10,000mg/L
Titration
Sulfate determination
100 packets sulfate reagent A (2 sets), 100mL LR sulfate reagent B, 100mL HR sulfate reagent B, 10mL sulfate reagent C, 20mL complexing agent, 30mL sulfate solution, 50mL plastic vessels (2), and 1mL syringes (2)
1 Kit
High range and low range determinations
Chemical Test Kit