missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hanna Instruments™ Chemical Test Kit
Includes all of the reagents and equipment necessary to perform manual water testing. Hanna Instruments™ Chemical Test Kit features pre-made, pre-measured reagents for fast, accurate determinations with high-resolution readings based on accepted titrations methods.
Brand: Hanna Instruments™ HI3830
This item is not returnable.
View return policy
Description
These chemical test kits use high quality reagents that are manufactured in Hanna's state-of-the-art facility and are clearly marked with the lot number and expiration date on each packet/bottle for traceability.
- Complete setup: All required materials are included, such as beakers, syringes, and premeasured reagent bottles and packets
- High resolution readings for fast, accurate determinations
- Replacement reagents available: No need to buy a new kit when reagents are exhausted
Specifications
DPD | |
Water samples | |
60 | |
0.0 to 3.0mg/L | |
Chemical Test Kit |
Bromine determination | |
30mL reagent 1, 20mL reagent 2, color comparison cube, and plastic vessel | |
1 Kit | |
Titration |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction