Learn More
Hanna Instruments™ Chemical Test Kit
Includes all of the reagents and equipment necessary to perform manual water testing. Hanna Instruments™ Chemical Test Kit features pre-made, pre-measured reagents for fast, accurate determinations with high-resolution readings based on accepted titrations methods.
Brand:  Hanna Instruments™ HI38018
 View more versions of this product
Product Code. 11692300
  • 1320.00 SEK / Each
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Description
These chemical test kits use high quality reagents that are manufactured in Hanna's state-of-the-art facility and are clearly marked with the lot number and expiration date on each packet/bottle for traceability.
  • Complete setup: All required materials are included, such as beakers, syringes, and premeasured reagent bottles and packets
  • High resolution readings for fast, accurate determinations
  • Replacement reagents available: No need to buy a new kit when reagents are exhausted
Specifications
Specifications
DPD
Water samples
100 ea. (200 total)
0.00 to 0.70mg/L and 0.0 to 3.5mg/L
Titration
Free chlorine LR/MR determination
HI93701-0 free chlorine reagent (200 packets), demineralizer bottle with cap for 12L, Checker disc, glass vials with caps (2), and 3mL plastic pipette
1 Kit
Low range and medium range determination
Chemical Test Kit
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • FREE CHLORINE LOW RANGE & MR TEST
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • EUH210-Safety data sheet available on request.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Purolite