Learn More
Heathrow Scientific Chest Freezer Rack, Stainless Steel
The Chest Freezer Rack safely stores samples in your freezer. Constructed of corrosion-resistant stainless steel racks that are compatible with major brands of chest freezers.
Brand: Heathrow Scientific HS2862DA
Additional Details : Weight : 1.00000kg
Description
- Chest Freezer Rack
- Rack has handle for easy transfer
- Robust mounting brackets assure handles stay securely in place for easy use
- Locking Rod Keeps tube storage boxes safely in place
Specifications
Chest Freezer Rack | |
Silver | |
Stainless Steel | |
Tube Boxes | |
5 | |
Box | |
5.5 in. | |
5.5 in. | |
16 in. |
5 x 1 | |
Tube Boxes | |
Non-sterile | |
HS2862DA | |
1 Ea. | |
14.1 cm | |
14.1 cm | |
40.6 cm |