Learn More
Fisherbrand™ Chest ULT Freezer Racks
Fisherbrand inventory control racks are ideal for all cold storage application. They fit all standard types of chest ULT freezers.
770.00 SEK - 830.00 SEK
Specifications
Product Type | Vertical Rack |
---|---|
Material | Stainless Steel |
For Use With (Application) | Standard 2 in. Cryoboxes |
For Use With (Equipment) | Chest ULT Freezer |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
15709539
|
Fisherbrand™
|
2 in. Boxes (13) |
830.00 SEK
Each |
Estimated Shipment: 08-10-2024
Log in to see stock. |
|||||
15779529
|
Fisherbrand™
|
2 in. Boxes (10) |
770.00 SEK
Each |
Estimated Shipment: 08-10-2024
Log in to see stock. |
|||||
15789529
|
Fisherbrand™
|
2 in. Boxes (11) |
795.00 SEK
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||||
15799529
|
Fisherbrand™
|
12 Boxes (2 in.) |
805.00 SEK
Each |
Estimated Shipment: 08-10-2024
Log in to see stock. |
|||||
Description
- Holds standard cryoboxes
- Made of high quality stainless steel
- Locking rod included
Specifications
Vertical Rack | |
Stainless Steel | |
Standard 2 in. Cryoboxes | |
Chest ULT Freezer |