Learn More
Thermo Scientific™ Chiller Plumbing Kits and Accessories
Choose these chiller plumbing kits and accessories, which include flexible tubing, tubing insulation and hose clamps.
Brand: Thermo Scientific™ 611000000107
Additional Details : Weight : 5.00000kg
Description
Thermo Scientific™ Chiller Plumbing Kits and accessories include flexible tubing, tubing insulation, and hose clamps.
Features:
- Include flexible tubing, tubing insulation, and hose clamps
Specifications
10m Plumbing Package | |
Tubing Kit | |
3/4 in. NPT |
ThermoFlex™ 3500 to 5000 with P3, P4 pumps | |
32.8 ft. (10 meters) of 5/8 in. ID Polybraid hose (use with 3/4 in. hose barbs) , insulation and clamps |