Learn More
Chloroacetone, 95%, stab. with 0.1% epoxidized soybean oil, Thermo Scientific Chemicals
679.00 SEK - 1680.00 SEK
Chemical Identifiers
CAS 78-95-5
Molecular Formula C3H5ClO
Molecular Weight (g/mol) 92.52
MDL Number MFCD00000936
InChI Key BULLHNJGPPOUOX-UHFFFAOYSA-N
SMILES CC(=O)CCl
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11471890
View Documents
Thermo Scientific Alfa Aesar
A11922.30
250 g
679.00 SEK
250g
11490980
View Documents
Thermo Scientific Alfa Aesar
A11922.0B
1000 g
1680.00 SEK
1000g
Description
Description
Chloroacetone was used in the synthesis of double-chain nonionic surfactants with an acid decomposition function via acid-catalyzed condensation with fatty alcohols (octyl, decyl and dodecyl). It was also used in the synthesis of meso-tetramethyl tetrakis-(4-phenoxy acetone)calix[4]pyrrole. It can also be used to make dye couplers for color photography and used in the Feist-Benary synthesis of furans.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Chloroacetone was used in the synthesis of double-chain nonionic surfactants with an acid decomposition function via acid-catalyzed condensation with fatty alcohols (octyl, decyl and dodecyl). It was also used in the synthesis of meso-tetramethyl tetrakis-(4-phenoxy acetone)calix[4]pyrrole. It can also be used to make dye couplers for color photography and used in the Feist-Benary synthesis of furans.
Solubility
Soluble in water (124g/L), ether and chloroform.
Notes
Refrigerate. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Do not store together with strong basic or oxidizing materials.
Specifications
Chemical Identifiers
78-95-5
92.52
BULLHNJGPPOUOX-UHFFFAOYSA-N
C3H5ClO
MFCD00000936
CC(=O)CCl
Specifications
78-95-5
1.161
40°C (104°F)
C3H5ClO
1.434
UN1695
14,2114
Soluble in water (124g/L),ether and chloroform.
CC(=O)CCl
92.52
92.53
Liquid
°C to 45°C
118°C to 120°C
95%, Stabilized with 0.1% Epoxidized Soybean Oil
CH2COCH3
MFCD00000936
605369
1-propanol, 3-chloro-, acetate, 3-chloropropanol acetate, 3-chlorprop-1-ylacetat, gamma-chloropropyl acetate, 3-chlorprop-1-ylacetat german, acetic acid 3-chloro-propyl ester, 1-acetoxy-3-chloropropane, .gamma.-chloropropyl acetate, 3-chloro-propyl acetate, acetic acid 3-chloropropyl ester
BULLHNJGPPOUOX-UHFFFAOYSA-N
3-chloropropyl acetate
12334
95%
Chloroacetone, Stabilized with 0.1% epoxidized soybean oil
Videos
Safety and Handling
Safety and Handling
GHS H Statement
H301-H310-H330-H314-H318-H226
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c
H226-H301+H311-H315-H319-H330-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: CHLOROACETONE, STABILIZED
missing translation for 'einecsNumber' : 201-161-1
missing translation for 'rtecsNumber' : UC0700000
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold