missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Cholic acid, 97% Product Code.: 10338943

Thermo Scientific Chemicals Cholic acid, 97%

Product Code. 10338943
500 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10338943

Brand: Thermo Scientific Chemicals 159115000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81-25-4
Molecular Formula C24H40O5
MDL Number MFCD00003672
InChI Key BHQCQFFYRZLCQQ-OELDTZBJSA-N
Synonym cholicacid, cholic acid, cholate, cholbam, 3alpha,7alpha,12alpha-trihydroxy-5beta-cholanic acid, cholsaeure, cholalin, cholic acid, 5beta, cholalic acid, colalinShow MoreShow Less
PubChem CID 221493
ChEBI CHEBI:16359

Specifications

Melting Point 197°C to 202°C
Color Beige to White
Loss on Drying 1% max. (135°C)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 500 g
Beilstein 10, IV, 2072
Merck Index 15, 2210
Specific Rotation (20°C, 589 nm) (c=2, ethanol)%gt; +35°
SMILES C[C@H](CCC(O)=O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C
IUPAC Name (4R)-4-[(1R,3aS,3bR,4R,5aS,7R,9aS,9bS,11S,11aR)-4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoic acid
Molecular Weight (g/mol) 408.58
Formula Weight 408.57
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material Cholic acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468352 Certificate TypeCertificate of Analysis Date19/12/2024 Catalog Number159115000,159110250,159111000
Lot NumberA0441861 Certificate TypeCertificate of Analysis Date20/05/2022 Catalog Number159110250,159115000,159111000
Lot NumberA0428228 Certificate TypeCertificate of Analysis Date18/03/2021 Catalog Number159110250,159115000,159111000
Lot NumberA0425339 Certificate TypeCertificate of Analysis Date14/12/2020 Catalog Number159110250,159115000,159111000
Lot NumberA0360991 Certificate TypeCertificate of Analysis Date18/06/2020 Catalog Number159110250,159115000,159111000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Cholic acid, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.